Assay Design



Gene Transcripts

Gene Symbol
tspan7
Entrez Gene ID
406802
Gene Name
tetraspanin 7
Gene Aliases
tm4sf2, wu:fa11g01, wu:fd18e11, wu:fl39g07, zgc:63823
Chromosome Location
Chr.14: 41469478 - 41478369 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214774.2 NP_999939.1 4 - 5 597 119
GenBank mRNA BC055255.1 - 4 - 5 537 119
BC097054.1 - 4 - 5 585 119
BC165114.1 - 4 - 5 469 119
BQ262702.1 - 4 - 5 505 119

View Full Product Details