Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214818.1 | NP_999983.1 | 20 - 21 | 2944 | 67 |
XM_005171284.4 | XP_005171341.1 | 19 - 20 | 2987 | 67 | |
XM_009298637.3 | XP_009296912.1 | 21 - 22 | 3051 | 67 | |
XM_021479667.1 | XP_021335342.1 | 20 - 21 | 3024 | 67 | |
GenBank mRNA | BC070002.1 | - | 20 - 21 | 2944 | 67 |