Assay Design



Gene Transcripts

Gene Symbol
psmc1b
Entrez Gene ID
415181
Gene Name
proteasome 26S subunit, ATPase 1b
Gene Aliases
wu:fj14c10, zgc:86923
Chromosome Location
Chr.20: 16721942 - 16740955 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002091.1 NP_001002091.1 5 - 6 499 83
GenBank mRNA BC071538.1 - 5 - 6 499 83

View Full Product Details