Assay Design



Gene Transcripts

Gene Symbol
gnb3a
Entrez Gene ID
436710
Gene Name
guanine nucleotide binding protein (G protein), beta polypeptide 3a
Gene Aliases
gnb3l, zgc:92641
Chromosome Location
Chr.16: 96513 - 123367 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002437.1 NP_001002437.1 3 - 4 447 73
XM_021466676.1 XP_021322351.1 3 - 4 371 73
GenBank mRNA BC076127.1 - 3 - 4 447 73

View Full Product Details