Assay Design



Gene Transcripts

Gene Symbol
grifin
Entrez Gene ID
445036
Gene Name
galectin-related inter-fiber protein
Gene Aliases
zgc:92897
Chromosome Location
Chr.3: 41785861 - 41791178 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003430.1 NP_001003430.1 3 - 4 297 87
GenBank mRNA BC076353.1 - 3 - 4 297 87

View Full Product Details