Assay Design



Gene Transcripts

Gene Symbol
st3gal8
Entrez Gene ID
445567
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 8
Gene Aliases
siat4, st3gal2l, zgc:111788
Chromosome Location
Chr.11: 26568490 - 26590450 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004017.1 NP_001004017.1 4 - 5 737 116
XM_009306027.3 XP_009304302.1 5 - 6 1293 116
XM_017358220.2 XP_017213709.1 4 - 5 518 116
GenBank mRNA AJ705061.1 - 3 - 4 413 116
AJ783740.1 - 4 - 5 737 116
BC093103.1 - 5 - 6 1250 116
BC164756.1 - 4 - 5 768 116

View Full Product Details