Assay Design



Gene Transcripts

Gene Symbol
gli2b
Entrez Gene ID
548610
Gene Name
GLI family zinc finger 2b
Gene Aliases
-
Chromosome Location
Chr.11: 43550194 - 43713750 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015069.1 NP_001015069.1 8 - 9 1426 73
GenBank mRNA AY928397.2 - 8 - 9 1426 73
BC162463.1 - 9 - 10 1223 73

View Full Product Details