Assay Design



Gene Transcripts

Gene Symbol
gm2a
Entrez Gene ID
550529
Gene Name
GM2 ganglioside activator
Gene Aliases
zgc:110188
Chromosome Location
Chr.14: 26224504 - 26228023 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017831.1 NP_001017831.1 2 - 3 244 70
XM_005173119.4 XP_005173176.1 2 - 3 281 70
GenBank mRNA BC092784.1 - 2 - 3 244 70
BC165264.1 - 1 - 2 135 70

View Full Product Details