Assay Design



Gene Transcripts

Gene Symbol
enpp4
Entrez Gene ID
550586
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4
Gene Aliases
E-NPP 4, zgc:113299
Chromosome Location
Chr.17: 80457 - 91957 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017887.1 NP_001017887.1 3 - 4 1202 78
GenBank mRNA BC093443.1 - 3 - 4 1202 78

View Full Product Details