Assay Design



Gene Transcripts

Gene Symbol
alg13
Entrez Gene ID
557181
Gene Name
ALG13 UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
otul
Chromosome Location
Chr.21: 34233496 - 34261789 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031838.1 NP_001027008.1 5 - 6 507 72
XM_005161414.4 XP_005161471.1 5 - 6 512 72
XM_005161417.4 XP_005161474.1 5 - 6 512 72
GenBank mRNA AY918066.1 - 5 - 6 507 72
BC057511.1 - 5 - 6 400 72
BC133108.1 - 5 - 6 426 72

View Full Product Details