Assay Design



Gene Transcripts

Gene Symbol
scn4aa
Entrez Gene ID
572442
Gene Name
sodium channel, voltage-gated, type IV, alpha, a
Gene Aliases
Na<sub>v</sub>1.4a, Nav1.4a
Chromosome Location
Chr.12: 51019 - 75228 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039825.1 NP_001034914.1 5 - 6 676 69
XM_017358479.2 XP_017213968.1 6 - 7 862 69
GenBank mRNA DQ149506.1 - 5 - 6 676 69

View Full Product Details