Assay Design



Gene Transcripts

Gene Symbol
edn2
Entrez Gene ID
569550
Gene Name
endothelin 2
Gene Aliases
si:ch211-202b2.2
Chromosome Location
Chr.19: 15276713 - 15281980 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045185.1 NP_001038650.1 1 - 2 128 72

View Full Product Details