Assay Design



Gene Transcripts

Gene Symbol
slc25a21
Entrez Gene ID
767694
Gene Name
solute carrier family 25 member 21
Gene Aliases
zgc:153387
Chromosome Location
Chr.17: 38030327 - 38233017 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076632.1 NP_001070100.1 2 - 3 141 71
GenBank mRNA BC124342.1 - 2 - 3 141 71

View Full Product Details