Assay Design



Gene Transcripts

Gene Symbol
mfge8b
Entrez Gene ID
558580
Gene Name
milk fat globule EGF and factor V/VIII domain containing b
Gene Aliases
mfge8, zgc:154157
Chromosome Location
Chr.25: 19149261 - 19192193 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080990.1 NP_001074459.1 4 - 5 511 62
XM_005163016.4 XP_005163073.1 4 - 5 576 62
GenBank mRNA BC131837.1 - 4 - 5 511 62

View Full Product Details