Assay Design



Gene Transcripts

Gene Symbol
zfr
Entrez Gene ID
321659
Gene Name
zinc finger RNA binding protein
Gene Aliases
hnrpa1, wu:fb25b08, zgc:66235, zgc:66403
Chromosome Location
Chr.5: 189463 - 207518 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199558.2 NP_955852.2 12 - 13 2171 65
GenBank mRNA BC055641.1 - - 1426 65
BC059198.1 - 12 - 13 2148 65

View Full Product Details