Assay Design
Gene Transcripts
- Gene Symbol
- zfr
- Entrez Gene ID
-
321659
- Gene Name
- zinc finger RNA binding protein
- Gene Aliases
-
hnrpa1, wu:fb25b08, zgc:66235, zgc:66403
- Chromosome Location
-
Chr.5: 189463 - 207518 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199558.2 |
NP_955852.2 |
4 - 5 |
591 |
61 |
GenBank mRNA |
BC059198.1 |
- |
4 - 5 |
568 |
61 |
View Full Product Details