Assay Design
Gene Transcripts
- Gene Symbol
- cox4i2
- Entrez Gene ID
-
393776
- Gene Name
- cytochrome c oxidase subunit 4I2
- Gene Aliases
-
zgc:73355
- Chromosome Location
-
Chr.23: 18944496 - 18951148 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200803.1 |
NP_957097.1 |
3 - 4 |
341 |
83 |
GenBank mRNA |
BC059669.1 |
- |
3 - 4 |
341 |
83 |
View Full Product Details