Assay Design



Gene Transcripts

Gene Symbol
irx2a
Entrez Gene ID
394032
Gene Name
iroquois homeobox 2a
Gene Aliases
Irx2, zgc:65872, zgc:77209, ziro2a
Chromosome Location
Chr.16: 558693 - 563174 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201057.1 NP_957351.1 2 - 3 761 119
GenBank mRNA AY845127.1 - 2 - 3 823 119
BC056322.1 - 2 - 3 761 119
BC065681.1 - 2 - 3 761 119

View Full Product Details