Assay Design
Gene Transcripts
- Gene Symbol
- cdc14aa
- Entrez Gene ID
-
394124
- Gene Name
- cell division cycle 14Aa
- Gene Aliases
-
CDC14A, zgc:63654
- Chromosome Location
-
Chr.2: 7075156 - 7105661 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_201149.1 |
NP_957443.1 |
14 - 15 |
2070 |
101 |
GenBank mRNA |
BC055188.1 |
- |
14 - 15 |
2070 |
101 |
View Full Product Details