Assay Design



Gene Transcripts

Gene Symbol
clic4
Entrez Gene ID
368255
Gene Name
chloride intracellular channel 4
Gene Aliases
cb142, wu:fb33c05, wu:fc30h10
Chromosome Location
Chr.17: 25444424 - 25480375 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201486.1 NP_958894.1 1 - 2 352 62
GenBank mRNA AY398328.1 - 1 - 2 332 62
BC051622.1 - 1 - 2 352 62
BC065609.1 - 1 - 2 267 62

View Full Product Details