Assay Design



Gene Transcripts

Gene Symbol
dgat1b
Entrez Gene ID
436731
Gene Name
diacylglycerol O-acyltransferase 1b
Gene Aliases
zgc:92327
Chromosome Location
Chr.16: 30948451 - 31000728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002458.1 NP_001002458.1 15 - 16 1526 94
XM_021466677.1 XP_021322352.1 14 - 15 1156 94
GenBank mRNA BC076012.1 - 15 - 16 1526 94

View Full Product Details