Assay Design



Gene Transcripts

Gene Symbol
cratb
Entrez Gene ID
447855
Gene Name
carnitine O-acetyltransferase b
Gene Aliases
crat, wu:fc39c02, zgc:92131
Chromosome Location
Chr.19: 18812805 - 18826746 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004594.2 NP_001004594.2 13 - 14 1750 67
GenBank mRNA BC081604.1 - 13 - 14 1677 67

View Full Product Details