Assay Design



Gene Transcripts

Gene Symbol
reep6
Entrez Gene ID
447918
Gene Name
receptor accessory protein 6
Gene Aliases
zgc:101529
Chromosome Location
Chr.11: 7261947 - 7272303 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004656.2 NP_001004656.1 6 - 7 712 81
GenBank mRNA BC081377.1 - 6 - 7 689 81
EH432211.1 - 6 - 7 744 81

View Full Product Details