Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012486.1 | NP_001012504.1 | 21 - 22 | 3001 | 73 |
XM_009306437.3 | XP_009304712.1 | 21 - 22 | 3400 | 73 | |
XM_009306438.3 | XP_009304713.1 | 20 - 21 | 3394 | 73 | |
XM_009306439.3 | XP_009304714.1 | 21 - 22 | 3400 | 73 | |
XM_009306440.3 | XP_009304715.1 | 21 - 22 | 3400 | 73 | |
GenBank mRNA | AY772391.1 | - | 21 - 22 | 3001 | 73 |