Assay Design



Gene Transcripts

Gene Symbol
spopla
Entrez Gene ID
541318
Gene Name
speckle type BTB/POZ protein like a
Gene Aliases
spopl, wu:fi71d08, zgc:101524
Chromosome Location
Chr.9: 10050890 - 10067947 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013447.1 NP_001013465.1 7 - 8 923 122
GenBank mRNA BC090815.1 - 7 - 8 923 122

View Full Product Details