Assay Design



Gene Transcripts

Gene Symbol
psma2
Entrez Gene ID
554153
Gene Name
proteasome 20S subunit alpha 2
Gene Aliases
wu:faa49c06, wu:fb98h03, zgc:110710
Chromosome Location
Chr.16: 43336570 - 43344870 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024441.1 NP_001019612.1 4 - 5 385 84
GenBank mRNA BC095382.1 - 4 - 5 385 84

View Full Product Details