Assay Design



Gene Transcripts

Gene Symbol
rtn1b
Entrez Gene ID
327065
Gene Name
reticulon 1b
Gene Aliases
rtn1, rtn1l, wu:fa21b09, wu:fj35h01
Chromosome Location
Chr.20: 20637760 - 20695953 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029948.2 NP_001025119.1 1 - 2 225 78
GenBank mRNA AY555036.1 - 1 - 2 214 78
BC052753.1 - 1 - 2 159 78
BC139604.1 - 1 - 2 159 78
BC153309.1 - 1 - 2 186 78

View Full Product Details