Assay Design



Gene Transcripts

Gene Symbol
mapre3a
Entrez Gene ID
559217
Gene Name
microtubule-associated protein, RP/EB family, member 3a
Gene Aliases
zgc:114065
Chromosome Location
Chr.17: 36866008 - 36896615 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030127.1 NP_001025298.1 5 - 6 656 95
XM_005158846.4 XP_005158903.1 5 - 6 762 95
GenBank mRNA BC097127.1 - 5 - 6 656 95

View Full Product Details