Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030183.1 | NP_001025354.1 | 7 - 8 | 1154 | 76 |
XM_005172625.4 | XP_005172682.1 | 8 - 9 | 1565 | 76 | |
XM_005172626.4 | XP_005172683.1 | 8 - 9 | 1565 | 76 | |
XM_005172627.4 | XP_005172684.1 | 8 - 9 | 1565 | 76 | |
XM_021479187.1 | XP_021334862.1 | 8 - 9 | 1565 | 76 | |
GenBank mRNA | BC163543.1 | - | 7 - 8 | 1223 | 76 |