Assay Design



Gene Transcripts

Gene Symbol
vgll2b
Entrez Gene ID
606659
Gene Name
vestigial-like family member 2b
Gene Aliases
zgc:110098
Chromosome Location
Chr.17: 27434626 - 27447527 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033095.1 NP_001028267.1 1 - 2 276 102
GenBank mRNA BC100003.1 - 1 - 2 276 102

View Full Product Details