Assay Design



Gene Transcripts

Gene Symbol
itgb1b
Entrez Gene ID
570216
Gene Name
integrin, beta 1b
Gene Aliases
cb810, itgb1
Chromosome Location
Chr.2: 43577293 - 43583881 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034987.1 NP_001030159.1 - 1459 72
GenBank mRNA BC162706.1 - - 1556 72
DQ149102.1 - - 1459 72

View Full Product Details