Assay Design



Gene Transcripts

Gene Symbol
reep3a
Entrez Gene ID
641577
Gene Name
receptor accessory protein 3a
Gene Aliases
fc41f04, reep3l, wu:fc41f04, zgc:123201
Chromosome Location
Chr.17: 19609698 - 19626299 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037575.1 NP_001032664.1 7 - 8 932 88
GenBank mRNA BC110095.1 - 7 - 8 932 88

View Full Product Details