Assay Design



Gene Transcripts

Gene Symbol
abcg2d
Entrez Gene ID
735310
Gene Name
ATP-binding cassette, sub-family G (WHITE), member 2d
Gene Aliases
-
Chromosome Location
Chr.1: 50564405 - 50592738 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042772.1 NP_001036237.1 8 - 9 1195 81
XM_009294223.3 XP_009292498.3 9 - 10 1551 81
GenBank mRNA DQ223861.1 - 8 - 9 1195 81

View Full Product Details