Assay Design



Gene Transcripts

Gene Symbol
abcg2a
Entrez Gene ID
735312
Gene Name
ATP-binding cassette, sub-family G (WHITE), member 2a
Gene Aliases
fd59c01, wu:fd59c01
Chromosome Location
Chr.23: 45909999 - 45936686 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042775.1 NP_001036240.1 2 - 3 249 63
XM_021469846.1 XP_021325521.1 2 - 3 283 63
GenBank mRNA DQ223858.1 - 2 - 3 249 63

View Full Product Details