Assay Design



Gene Transcripts

Gene Symbol
jph1b
Entrez Gene ID
556485
Gene Name
junctophilin 1b
Gene Aliases
zgc:136916
Chromosome Location
Chr.2: 30290454 - 30324325 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044348.1 NP_001037813.1 5 - 6 2305 150
GenBank mRNA BC117663.1 - 5 - 6 2305 150

View Full Product Details