Assay Design



Gene Transcripts

Gene Symbol
sall4
Entrez Gene ID
572527
Gene Name
spalt-like transcription factor 4
Gene Aliases
cb372, cb614, ik:tdsubc_2c1, sb:cb372, sb:cb614, wu:fa01g03, xx:tdsubc_2c1
Chromosome Location
Chr.23: 38941361 - 38949912 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080609.1 NP_001074078.1 1 - 2 103 105
GenBank mRNA EF051583.1 - 1 - 2 103 105

View Full Product Details