Assay Design



Gene Transcripts

Gene Symbol
hspa13
Entrez Gene ID
560481
Gene Name
heat shock protein 70 family, member 13
Gene Aliases
stch, zgc:162281
Chromosome Location
Chr.15: 29549115 - 29556698 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089479.1 NP_001082948.1 3 - 4 636 81
GenBank mRNA BC139562.1 - 3 - 4 636 81

View Full Product Details