Assay Design



Gene Transcripts

Gene Symbol
wnt11
Entrez Gene ID
30283
Gene Name
wingless-type MMTV integration site family, member 11
Gene Aliases
fk73h04, wnt11f1, wnt11r, wu:fk73h04
Chromosome Location
Chr.10: 32104343 - 32123204 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131076.2 NP_571151.1 2 - 3 409 67
GenBank mRNA AF249266.1 - 2 - 3 323 67
BC066498.1 - 2 - 3 409 67
KM655705.1 - 1 - 2 89 67

View Full Product Details