Assay Design



Gene Transcripts

Gene Symbol
ndrg3a
Entrez Gene ID
30286
Gene Name
ndrg family member 3a
Gene Aliases
fc04h03, ndr3, ndrg3, wu:fc04h03, zgc:63551
Chromosome Location
Chr.11: 25064499 - 25139061 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199517.1 NP_955811.1 13 - 14 975 88
XM_005169249.3 XP_005169306.1 13 - 14 1007 88
XM_005169250.4 XP_005169307.1 11 - 12 1043 88
XM_009306019.3 XP_009304294.1 13 - 14 1012 88
XM_021479547.1 XP_021335222.1 13 - 14 1029 88
GenBank mRNA AY398386.1 - 13 - 14 1017 88
BC055138.1 - 13 - 14 975 88

View Full Product Details