Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199795.2 | NP_956089.1 | 2 - 3 | 368 | 77 |
GenBank mRNA | AY398374.1 | - | 2 - 3 | 349 | 77 |
AY398431.1 | - | 2 - 3 | 368 | 77 | |
BC046889.1 | - | 2 - 3 | 319 | 77 | |
BC066482.1 | - | 2 - 3 | 347 | 77 | |
BC164535.1 | - | 1 - 2 | 256 | 77 |