Assay Design



Gene Transcripts

Gene Symbol
chp2
Entrez Gene ID
327599
Gene Name
calcineurin-like EF-hand protein 2
Gene Aliases
wu:fi19e07, zgc:55363
Chromosome Location
Chr.15: 313376 - 319535 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199836.2 NP_956130.1 1 - 2 261 63
GenBank mRNA BC044149.1 - 1 - 2 262 63
BC163944.1 - 1 - 2 105 63

View Full Product Details