Assay Design



Gene Transcripts

Gene Symbol
chp2
Entrez Gene ID
327599
Gene Name
calcineurin-like EF-hand protein 2
Gene Aliases
wu:fi19e07, zgc:55363
Chromosome Location
Chr.15: 313376 - 319535 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199836.2 NP_956130.1 2 - 3 338 85
GenBank mRNA BC044149.1 - 2 - 3 339 85
BC163944.1 - 2 - 3 182 85

View Full Product Details