Assay Design
Gene Transcripts
- Gene Symbol
- cldn15a
- Entrez Gene ID
-
573644
- Gene Name
- claudin 15a
- Gene Aliases
-
cldn15, cldn15l, zgc:63943
- Chromosome Location
-
Chr.7: 20830762 - 20836787 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200404.1 |
NP_956698.1 |
3 - 4 |
624 |
89 |
GenBank mRNA |
BC054577.1 |
- |
3 - 4 |
624 |
89 |
View Full Product Details