Assay Design



Gene Transcripts

Gene Symbol
cldn15a
Entrez Gene ID
573644
Gene Name
claudin 15a
Gene Aliases
cldn15, cldn15l, zgc:63943
Chromosome Location
Chr.7: 20830762 - 20836787 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200404.1 NP_956698.1 4 - 5 734 78
GenBank mRNA BC054577.1 - 4 - 5 734 78
BC153629.1 - 4 - 5 740 78

View Full Product Details