Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_200427.2 | NP_956721.2 | 1 - 2 | 581 | 100 |
GenBank mRNA | AF411696.1 | - | 1 - 2 | 497 | 100 |
BC054674.1 | - | 1 - 2 | 579 | 100 | |
BC165014.1 | - | 1 - 2 | 470 | 100 | |
CF996038.1 | - | 1 - 2 | 145 | 100 | |
DT068839.1 | - | 1 - 2 | 146 | 100 |