Assay Design



Gene Transcripts

Gene Symbol
ap2m1b
Entrez Gene ID
394001
Gene Name
adaptor related protein complex 2 subunit mu 1b
Gene Aliases
ap2m1, zgc:56643
Chromosome Location
Chr.18: 45954991 - 45984589 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201026.2 NP_957320.1 5 - 6 746 101
XM_005159256.4 XP_005159313.1 6 - 7 797 101
XM_009293860.3 XP_009292135.1 6 - 7 797 101
GenBank mRNA BC049515.1 - 5 - 6 748 101

View Full Product Details