Assay Design



Gene Transcripts

Gene Symbol
rhag
Entrez Gene ID
405771
Gene Name
Rh associated glycoprotein
Gene Aliases
wu:fj61d12
Chromosome Location
Chr.20: 14016286 - 14054133 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212845.1 NP_998010.1 4 - 5 682 103
XM_009294595.3 XP_009292870.1 4 - 5 732 103
GenBank mRNA AF531094.1 - 4 - 5 682 103
BC159236.1 - 4 - 5 690 103
BC162203.1 - 4 - 5 680 103
BC162529.1 - 4 - 5 680 103

View Full Product Details