Assay Design



Gene Transcripts

Gene Symbol
atp13a1
Entrez Gene ID
334466
Gene Name
ATPase 13A1
Gene Aliases
atp13a, hm:zehn0386, wu:fi75c10, zgc:63781
Chromosome Location
Chr.3: 52899801 - 52952026 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001403.2 NP_001001403.2 19 - 20 2841 66
GenBank mRNA BC055391.1 - 19 - 20 2841 66

View Full Product Details