Assay Design



Gene Transcripts

Gene Symbol
slc1a4
Entrez Gene ID
368885
Gene Name
solute carrier family 1 member 4
Gene Aliases
si:zc101n13.5, zgc:92817
Chromosome Location
Chr.13: 24431213 - 24447540 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002513.2 NP_001002513.2 2 - 3 561 98

View Full Product Details