Assay Design



Gene Transcripts

Gene Symbol
tnfsf10
Entrez Gene ID
436866
Gene Name
TNF superfamily member 10
Gene Aliases
tnfsf10l2, zgc:92320
Chromosome Location
Chr.24: 26932472 - 26937526 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002593.2 NP_001002593.1 3 - 4 383 108
GenBank mRNA BC076005.1 - 3 - 4 334 108
EH448113.1 - 3 - 4 376 108

View Full Product Details